6-Methyl-5-hepten-2-one 98%

Supplier: Ambeed
Danger

A565531-25G A565531-100G
76774-974EA 14 USD
76774-974 77627-164
6-Methyl-5-hepten-2-one 98%
6-Methyl-5-hepten-2-one

Precaución: for Research Use Only

Formula: C₈H₁₄O
MDL Number: MFCD00008905
CAS Number: 110-93-0
UN: 1224
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR