6-Methyl-5-nitroquinoline 98%

Supplier: Ambeed
Danger

A564251-250MG A564251-25G A564251-5G
76774-968EA 7.43 USD
76774-968 76774-970 76774-972
6-Methyl-5-nitroquinoline 98%
6-Methyl-5-nitroquinoline

Precaución: for Research Use Only

Formula: C₁₀H₈N₂O₂
MW: 188.19 g/mol
MDL Number: MFCD00024021
CAS Number: 23141-61-9
UN: 2923
ADR: 8,6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR