2'-Hydroxy-4',6'-dimethoxyacetophenone 95%
Supplier: Ambeed
|
Warning
|
A564222-100MG
A564222-10MG
A564222-250MG
A564222-25G
A564222-25MG
A564222-50MG
A564222-5G
A564222-1G
A564222-100G
76774-954EA
13
USD
76774-954
76774-956
76774-958
76774-960
76774-962
76774-964
76774-966
77598-574
77878-362
2'-Hydroxy-4',6'-dimethoxyacetophenone 95%
2'-Hydroxy-4',6'-dimethoxyacetophenone
Precaución: for Research Use Only
Formula:
C₁₀H₁₂O₄ |
MDL Number:
MFCD00017243 CAS Number: 90-24-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...