3-Chloro-5-nitrobenzotrifluoride ≥95% (by GC)

Supplier: Ambeed
Warning

Synonyms: 1-Chloro-3-nitro-5-(trifluoromethyl)benzene

A562947-10G A562947-1G A562947-5G
76774-936EA 136.86 USD
76774-936 76774-938 76774-940
3-Chloro-5-nitrobenzotrifluoride ≥95% (by GC)
3-Chloro-5-nitrobenzotrifluoride

Precaución: for Research Use Only

Formula: C₇H₃ClF₃NO₂
MW: 225.5500 g/mol
MDL Number: MFCD03412202
CAS Number: 401-93-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR