5-Dihydroxyboronylnicotinic acid 98%

Supplier: Ambeed
Warning

Synonyms: 5-Carboxypyridine-3-boronic acid

A562873-1G A562873-250MG A562873-5G
76774-930EA 202.64 USD
76774-930 76774-932 76774-934
5-Dihydroxyboronylnicotinic acid 98%
5-Dihydroxyboronylnicotinic acid

Precaución: for Research Use Only

Formula: C₆H₆BNO₄
MW: 166.93 g/mol
MDL Number: MFCD02323212
CAS Number: 913836-03-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR