5-Phenyl-1H-indole 95%

Supplier: Ambeed
Danger

A562458-100MG A562458-1G A562458-250MG
76774-924EA 119.89 USD
76774-924 76774-926 76774-928
5-Phenyl-1H-indole 95%
5-Phenyl-1H-indole

Precaución: for Research Use Only

Formula: C₁₄H₁₁N
MW: 193.244 g/mol
MDL Number: MFCD06801971
CAS Number: 66616-72-6
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR