6-Acetyl-2H-1,4-benzoxazin-3(4H)-one 98%

Supplier: Ambeed
Warning

A560468-1G A560468-25G A560468-5G
76774-910EA 6 USD
76774-910 76774-912 76774-914
6-Acetyl-2H-1,4-benzoxazin-3(4H)-one 98%
6-Acetyl-2H-1,4-benzoxazin-3(4H)-one

Precaución: for Research Use Only

Formula: C₁₀H₉NO₃
MDL Number: MFCD01321313
CAS Number: 26518-71-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR