5-Methylisoxazole-4-sulfonyl chloride 97%

Supplier: Ambeed
Danger

Synonyms: 5-Methyl-1,2-oxazole-4-sulphonyl chloride, 4-(Chlorosulphonyl)-5-methylisoxazole, 5-Methyl-4-isoxazolesulphonyl chloride

A559273-1G A559273-5G A559273-250MG
76774-908EA 48.81 USD
76774-908 77572-764 77598-568
5-Methylisoxazole-4-sulfonyl chloride 97%
5-Methylisoxazole-4-sulfonyl chloride

Precaución: for Research Use Only

Formula: C₄H₄ClNO₃S
MW: 181.597 g/mol
MDL Number: MFCD02681975
CAS Number: 321309-26-6
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR