6-Methoxy-2-phenyl-tetralone 97%

Supplier: Ambeed
Warning

Synonyms: 6-methoxy-2-phenyl-3,4-dihydro-2H-naphthalen-1-one

A557346-1G A557346-250MG
76774-892EA 38 USD
76774-892 76774-894
6-Methoxy-2-phenyl-tetralone 97%
6-Methoxy-2-phenyl-tetralone

Precaución: for Research Use Only

Formula: C₁₇H₁₆O₂
MDL Number: MFCD10698841
CAS Number: 1769-84-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR