6-(Fmoc-amino)-1-hexanol 97%

Supplier: Ambeed
Warning

Synonyms: 9-Fluorenylmethyl N-(6-hydroxyhexyl)carbamate, (9H-Fluoren-9-yl)methyl (6-hydroxyhexyl)carbamate, 6-Fmoc-Acp-ol

A554141-1G A554141-250MG A554141-5G
76774-872EA 15 USD
76774-872 76774-874 77627-142
6-(Fmoc-amino)-1-hexanol 97%
6-(Fmoc-amino)-1-hexanol

Precaución: for Research Use Only

Formula: C₂₁H₂₅NO₃
MDL Number: MFCD00380196
CAS Number: 127903-20-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR