6-Hydroxy-7-methoxy-2H-chromen-2-one 98+%

Supplier: Ambeed
Warning

A554035-100MG A554035-50MG
76774-870EA 154 USD
76774-870 77878-342
6-Hydroxy-7-methoxy-2H-chromen-2-one 98+%
6-Hydroxy-7-methoxy-2H-chromen-2-one

Precaución: for Research Use Only

Formula: C₁₀H₈O₄
MDL Number: MFCD00016976
CAS Number: 776-86-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR