EMCA (6-Maleimidohexanoic acid) 98%

Supplier: Ambeed
Warning

Synonyms: N-Maleoyl-6-aminocaproic acid, ε-Maleimidocaproic acid, 2,5-Dihydro-2,5-dioxo-1H-pyrrole-1-hexanoic acid, 6-Maleimide hexanoic acid, N-(5-Carboxypentyl)maleimide, MCA, Maleimidocaproic acid, 6-Maleimidohexanoic acid, 6-Maleimidocaproic acid

A553874-100G A553874-10G A553874-1G A553874-25G A553874-500G A553874-5G
76774-858EA 100 USD
76774-858 76774-860 76774-862 76774-864 76774-866 76774-868
EMCA (6-Maleimidohexanoic acid) 98%
EMCA (6-Maleimidohexanoic acid)

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₄
MW: 211.22 g/mol
MDL Number: MFCD00043140
CAS Number: 55750-53-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR