Ethyl diacetoacetate 95%

Supplier: Ambeed
Warning

Synonyms: Diacetoacetic Acid Ethyl Ester, Ethyl 2-Acetyl-3-oxobutyrate, Ethyl 2-acetyl-3-oxobutanoate

A553864-100G A553864-1G A553864-25G A553864-5G
76774-850EA 202.64 USD
76774-850 76774-852 76774-854 76774-856
Ethyl diacetoacetate 95%
Ethyl diacetoacetate

Precaución: for Research Use Only

Formula: C₈H₁₂O₄
MW: 172.18 g/mol
MDL Number: MFCD00009864
CAS Number: 603-69-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR