6-Chloro-2H-[1,4]benzoxazine-3(4H)-one 98%

Supplier: Ambeed
Warning

A552041-1G A552041-25G A552041-5G
76774-844EA 10 USD
76774-844 76774-846 76774-848
6-Chloro-2H-[1,4]benzoxazine-3(4H)-one 98%
6-Chloro-2H-[1,4]benzoxazine-3(4H)-one

Precaución: for Research Use Only

Formula: C₈H₆ClNO₂
MDL Number: MFCD00174511
CAS Number: 7652-29-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR