5,6-Dihydro-2H-pyran-2-one 95%

Supplier: Ambeed
Danger

A550479-10G A550479-1G A550479-250MG A550479-25G A550479-5G
76774-832EA 217 USD
76774-832 76774-834 76774-836 77627-134 77627-136
5,6-Dihydro-2H-pyran-2-one 95%
5,6-Dihydro-2H-pyran-2-one

Precaución: for Research Use Only

Formula: C₅H₆O₂
MDL Number: MFCD00010439
CAS Number: 3393-45-1
UN: 1224
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR