5-Acetyl-2,4-dimethylthiazole 98%

Supplier: Ambeed
Warning

Synonyms: 2,4-Dimethyl-5-acetylthiazole, 1-(2,4-Dimethylthiazol-5-yl)ethanone, 1-(2,4-Dimethylthiazol-5-yl)ethan-1-one

A547786-10G A547786-1G A547786-5G
76774-808EA 9 USD
76774-808 76774-810 76774-812
5-Acetyl-2,4-dimethylthiazole 98%
5-Acetyl-2,4-dimethylthiazole

Precaución: for Research Use Only

Formula: C₇H₉NOS
MW: 155.22 g/mol
Boiling Pt: 228
MDL Number: MFCD00005331
CAS Number: 38205-60-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR