Methyl 5-bromo-2-nitrobenzoate 98%

Supplier: Ambeed
Warning

A544133-10G A544133-1G A544133-25G A544133-5G
76774-780EA 45 USD
76774-780 76774-782 76774-784 76774-786
Methyl 5-bromo-2-nitrobenzoate 98%
Methyl 5-bromo-2-nitrobenzoate

Precaución: for Research Use Only

Formula: C₈H₆BrNO₄
MDL Number: MFCD00092886
CAS Number: 883554-93-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR