5,10,15,20-Tetrakis(pentafluorophenyl)porphyrin 98%

Supplier: Ambeed
Warning

Synonyms: 5,10,15,20-Tetrakis(perfluorophenyl)porphyrin, 5,10,15,20-Tetrakis(pentafluorophenyl)-21H,23H-porphine, 5,10,15,20-Tetrakis(pentafluorophenyl)-21h,23h-porphine

A543645-100MG A543645-1G A543645-250MG
76774-774EA 62 USD
76774-774 76774-776 76774-778
5,10,15,20-Tetrakis(pentafluorophenyl)porphyrin 98%
5,10,15,20-Tetrakis(pentafluorophenyl)porphyrin

Precaución: for Research Use Only

Formula: C₄₄H₁₀F₂₀N₄
MW: 974.56 g/mol
MDL Number: MFCD00010032
CAS Number: 25440-14-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR