1-Isopropyl-4-(2-nitrovinyl)benzene 95%

Supplier: Ambeed
Warning

A542740-10G A542740-5G
76774-770EA 267.35 USD
76774-770 76774-772
1-Isopropyl-4-(2-nitrovinyl)benzene 95%
1-Isopropyl-4-(2-nitrovinyl)benzene

Precaución: for Research Use Only

Formula: C₁₁H₁₃NO₂
MW: 191.22 g/mol
MDL Number: MFCD00085100
CAS Number: 42139-37-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR