Ethyl-3,4,5-rimethoxybenzoate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 3,4,5-trimethoxybenzoate, 3,4,5-Trimethoxybenzoic acid ethyl ester

A541743-10G A541743-1G A541743-1MG A541743-25G A541743-5G A541743-100G
76774-760EA 87 USD
76774-760 76774-762 76774-764 76774-766 76774-768 77627-116
Ethyl-3,4,5-rimethoxybenzoate 98%
Ethyl-3,4,5-rimethoxybenzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₅
MW: 240.26 g/mol
MDL Number: MFCD00017525
CAS Number: 6178-44-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR