(±)-γ-Decalactone 96%

Supplier: Ambeed
Warning

A537002-25G A537002-5G A537002-1000G A537002-100G A537002-500G
76774-756EA 8 USD
76774-756 76774-758 77627-094 77627-096 77627-098
(±)-γ-Decalactone 96%
(±)-γ-Decalactone

Precaución: for Research Use Only

Formula: C₁₀H₁₈O₂
MDL Number: MFCD00005404
CAS Number: 706-14-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR