5-Chloro-2-(methylsulfonyl)pyrimidine-4-carboxylic acid 99+%

Supplier: Ambeed
Warning

A536947-10MG A536947-1MG A536947-25MG A536947-5MG
76774-748EA 146.41 USD
76774-748 76774-750 76774-752 76774-754
5-Chloro-2-(methylsulfonyl)pyrimidine-4-carboxylic acid 99+%
5-Chloro-2-(methylsulfonyl)pyrimidine-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₅ClN₂O₄S
MW: 236.63 g/mol
MDL Number: MFCD00507244
CAS Number: 38275-34-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR