Zinc(II) (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate 98%

Supplier: Ambeed
Warning

Synonyms: Zinc Gluconate

A536852-100G A536852-500G A536852-1000G
76774-744EA 6 USD
76774-744 76774-746 77627-092
Zinc(II) (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate 98%
Zinc(II) (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate

Precaución: for Research Use Only

Formula: C₁₂H₂₂O₁₄Zn
MW: 455.70 g/mol
MDL Number: MFCD04037230
CAS Number: 4468-02-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR