5-Chlorothiophene-2-sulfonohydrazide 95%

Supplier: Ambeed
Danger

A536457-100MG A536457-1G A536457-250MG
76774-738EA 62 USD
76774-738 76774-740 76774-742
5-Chlorothiophene-2-sulfonohydrazide 95%
5-Chlorothiophene-2-sulfonohydrazide

Precaución: for Research Use Only

Formula: C₄H₅ClN₂O₂S₂
MDL Number: MFCD00052583
CAS Number: 78380-28-6
UN: 1325
ADR: 4.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR