4-Iodo-2-methylphenol 95%

Supplier: Ambeed
Warning

A536014-1G A536014-250MG A536014-25G A536014-5G
76774-730EA 30.77 USD
76774-730 76774-732 76774-734 76774-736
4-Iodo-2-methylphenol 95%
4-Iodo-2-methylphenol

Precaución: for Research Use Only

Formula: C₇H₇IO
MW: 234.03 g/mol
MDL Number: MFCD00075092
CAS Number: 60577-30-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR