6-Chloro-3-hydroxypyridazine-4-carboxylic acid 98%

Supplier: Ambeed
Warning

A533064-100MG A533064-1G A533064-250MG
76774-718EA 41.38 USD
76774-718 76774-720 76774-722
6-Chloro-3-hydroxypyridazine-4-carboxylic acid 98%
6-Chloro-3-hydroxypyridazine-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₃ClN₂O₃
MW: 174.5419 g/mol
MDL Number: MFCD12756225
CAS Number: 50681-26-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR