Chromone-2-carboxylic acid 98%

Supplier: Ambeed
Warning

A526233-10G A526233-1G A526233-250MG A526233-25G A526233-25MG A526233-5G A526233-100G
76774-684EA 28 USD
76774-684 76774-686 76774-688 76774-690 76774-692 76774-694 77627-074
Chromone-2-carboxylic acid 98%
Chromone-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₆O₄
MDL Number: MFCD00006838
CAS Number: 4940-39-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR