Methyl-4-Isopropoxybenzoate 95%

Supplier: Ambeed
Warning

Synonyms: 4-Isopropoxybenzoic Acid Methyl Ester, Methyl 4-isopropoxybenzoate

A523943-10G A523943-1G A523943-5G
76774-658EA 82.76 USD
76774-658 76774-660 76774-662
Methyl-4-Isopropoxybenzoate 95%
Methyl-4-Isopropoxybenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₃
MW: 194.23 g/mol
MDL Number: MFCD00084855
CAS Number: 35826-59-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR