[Bicyclo[2.2.1]hept-5-en-2-yl]triethoxysilane (mixture of isomers) 97%

Supplier: Ambeed
Warning

Synonyms: 5-(Triethoxysilyl)-2-norbornene, 5-(Triethoxysilyl)bicyclo[2.2.1]hept-2-ene, Bicyclo[2.2.1]hept-5-en-2-yltriethoxysilane

A523805-25G A523805-5G
76774-654EA 176.11 USD
76774-654 76774-656
[Bicyclo[2.2.1]hept-5-en-2-yl]triethoxysilane (mixture of isomers) 97%
[Bicyclo[2.2.1]hept-5-en-2-yl]triethoxysilane (mixture of isomers)

Precaución: for Research Use Only

Formula: C₁₃H₂₄O₃Si
MW: 256.42 g/mol
MDL Number: MFCD00167591
CAS Number: 18401-43-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR