5,7-Dimethoxycoumarin 95%

Supplier: Ambeed
Warning

Synonyms: Citropten, 5,7-Dimethoxy-2H-chromen-2-one

A520613-1G A520613-1MG A520613-250MG A520613-25MG A520613-50MG
76774-638EA 185.66 USD
76774-638 76774-640 76774-642 76774-644 76774-646
5,7-Dimethoxycoumarin 95%
5,7-Dimethoxycoumarin

Precaución: for Research Use Only

Formula: C₁₁H₁₀O₄
MW: 206.20 g/mol
MDL Number: MFCD00006870
CAS Number: 487-06-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR