α,α-Dimethylpropiophenone 97%

Supplier: Ambeed
Warning

Synonyms: 2,2,2-Trimethylacetophenone

A518255-1G A518255-250MG A518255-5G
76774-632EA 51.99 USD
76774-632 76774-634 77572-648
α,α-Dimethylpropiophenone 97%
α,α-Dimethylpropiophenone

Precaución: for Research Use Only

Formula: C₁₁H₁₄O
MW: 162.23 g/mol
MDL Number: MFCD00008844
CAS Number: 938-16-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR