5-Hexyl-2,2'-bithiophene 98+%

Supplier: Ambeed
Danger

A516334-100MG A516334-1G A516334-250MG
76774-620EA 10.61 USD
76774-620 76774-622 76774-624
5-Hexyl-2,2'-bithiophene 98+%
5-Hexyl-2,2'-bithiophene

Precaución: for Research Use Only

Formula: C₁₄H₁₈S₂
MW: 250.42 g/mol
MDL Number: MFCD05664373
CAS Number: 173448-31-2
UN: 2922
ADR: 8,6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR