3,5-Dimethyl-p-anisic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3,5-Dimethyl-p-anisic acid, 4-Methoxy-3,5-dimethylbenzoic acid, 3,5-Dimethyl-4-methoxybenzenecarboxylic acid, 3,5-Dimethyl-4-methoxybenzoic acid

A515637-10G A515637-1G A515637-250MG A515637-25G A515637-5G
76774-602EA 48.81 USD
76774-602 76774-604 76774-606 76774-608 76774-610
3,5-Dimethyl-p-anisic acid 98%
3,5-Dimethyl-p-anisic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₃
MW: 180.2 g/mol
MDL Number: MFCD00020309
CAS Number: 21553-46-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR