6-Nitro-1,3-benzoxazol-2(3H)-one 98%

Supplier: Ambeed
Warning

Synonyms: 6-Nitrobenzoxazole-2(3H)-one

A513955-100G A513955-1G A513955-25G A513955-5G
76774-590EA 395 USD
76774-590 76774-592 76774-594 76774-596
6-Nitro-1,3-benzoxazol-2(3H)-one 98%
6-Nitro-1,3-benzoxazol-2(3H)-one

Precaución: for Research Use Only

Formula: C₇H₄N₂O₄
MDL Number: MFCD00463755
CAS Number: 4694-91-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR