Phenyl(phenylamino)acetic acid 97%

Supplier: Ambeed
Warning

Synonyms: Anilino(phenyl)acetic acid, Phenyl(phenylamino)ethanoic acid

A513452-100MG A513452-250MG
76774-586EA 49.87 USD
76774-586 76774-588
Phenyl(phenylamino)acetic acid 97%
Phenyl(phenylamino)acetic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₃NO₂
MW: 227.259 g/mol
MDL Number: MFCD01724775
CAS Number: 3684-12-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR