5-(Methoxycarbonyl)furan-2-boronic acid 95%

Supplier: Ambeed
Warning

Synonyms: (5-(Methoxycarbonyl)furan-2-yl)boronic acid

A512873-100MG A512873-1G A512873-250MG A512873-5G
76774-578EA 27 USD
76774-578 76774-580 76774-582 76774-584
5-(Methoxycarbonyl)furan-2-boronic acid 95%
5-(Methoxycarbonyl)furan-2-boronic acid

Precaución: for Research Use Only

Formula: C₆H₇BO₅
MW: 169.928 g/mol
MDL Number: MFCD16660203
CAS Number: 876189-20-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR