α,α,α,α',α',α'-Hexafluoro-5-nitroxylene 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-Nitro-3,5-bis(trifluoromethyl)benzene
A510763-1G
A510763-100G
A510763-25G
A510763-5G
76774-568EA
8.49
USD
76774-568
77551-532
77551-534
77602-192
α,α,α,α',α',α'-Hexafluoro-5-nitroxylene 98%
α,α,α,α',α',α'-Hexafluoro-5-nitroxylene
Precaución: for Research Use Only
Formula:
C₈H₃F₆NO₂ MW: 259.11 g/mol |
MDL Number:
MFCD00000384 CAS Number: 328-75-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...