5-Amino-6-methylquinoline 98+%

Supplier: Ambeed
Danger

Synonyms: 6-Methylquinolin-5-amine

A510338-100MG A510338-1G A510338-250MG A510338-5G
76774-560EA 43.5 USD
76774-560 76774-562 76774-564 76774-566
5-Amino-6-methylquinoline 98+%
5-Amino-6-methylquinoline

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂
MW: 158.1998 g/mol
MDL Number: MFCD00094194
CAS Number: 50358-35-5
UN: 2923
ADR: 8,6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR