Methyl-5-bromobenzo[b]thiophene-2-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 5-bromobenzo[b]thiophene-2-carboxylate, Methyl 5-bromo-1-benzothiophene-2-carboxylate

A510059-5G A510059-1G
76774-558EA 197.33 USD
76774-558 77627-010
Methyl-5-bromobenzo[b]thiophene-2-carboxylate 98%
Methyl 5-bromobenzo[b]thiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₇BrO₂S
MW: 271.1300 g/mol
MDL Number: MFCD05662034
CAS Number: 7312-11-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR