5-Fluoro-1-benzofuran-2-carboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: 5-Fluorobenzofuran-2-carboxylic acid

A509826-1G A509826-5G A509826-250MG
76774-554EA 75.33 USD
76774-554 76774-556 77627-004
5-Fluoro-1-benzofuran-2-carboxylic acid 95%
5-Fluoro-1-benzofuran-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₅FO₃
MW: 180.133 g/mol
MDL Number: MFCD09038172
CAS Number: 89197-62-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR