2',3',6'-Trifluoroacetophenone 97%

Supplier: Ambeed
Warning

A506915-1G A506915-5G A506915-25G A506915-100G
76774-546EA 15 USD
76774-546 76774-548 77551-526 77568-990
2',3',6'-Trifluoroacetophenone 97%
2',3',6'-Trifluoroacetophenone

Precaución: For research use only.

Formula: C₈H₅F₃O
MDL Number: MFCD00070809
CAS Number: 208173-22-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR