4-Fluorophthalic anhydride 97%

Supplier: Ambeed
Warning

Synonyms: 5-Fluoroisobenzofuran-1,3-dione

A504742-1G A504742-250MG A504742-25G A504742-5G A504742-100G
76774-522EA 12 USD
76774-522 76774-524 76774-526 76774-528 77878-288
4-Fluorophthalic anhydride 97%
4-Fluorophthalic anhydride

Precaución: for Research Use Only

Formula: C₈H₃FO₃
MDL Number: MFCD00191363
CAS Number: 319-03-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR