4-Methoxy-3,6-dihydro-2H-pyran 95%

Supplier: Ambeed
Danger

Synonyms: 5,6-Dihydro-4-methoxy-2H-pyran

A503517-100MG A503517-250MG
76774-506EA 44.56 USD
76774-506 76774-508
4-Methoxy-3,6-dihydro-2H-pyran 95%
4-Methoxy-3,6-dihydro-2H-pyran

Precaución: for Research Use Only

Formula: C₆H₁₀O₂
MW: 114.15 g/mol
MDL Number: MFCD00006570
CAS Number: 17327-22-9
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR