4-(Boc-amino)-1-Fmoc-piperidine-4-carboxylic Acid 95%
Supplier: Ambeed
|
Warning
|
A502185-100MG
A502185-1G
A502185-250MG
A502185-25G
A502185-5G
76774-492EA
20.16
USD
76774-492
76774-494
76774-496
76774-498
77594-572
4-(Boc-amino)-1-Fmoc-piperidine-4-carboxylic Acid 95%
4-(Boc-amino)-1-Fmoc-piperidine-4-carboxylic Acid
Precaución: for Research Use Only
Formula:
C₂₆H₃₀N₂O₆ MW: 466.53 g/mol |
MDL Number:
MFCD02683117 CAS Number: 368866-07-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...