3',5'-Difluoro-2'-hydroxyacetophenone 98%

Supplier: Ambeed
Warning

Synonyms: 1-(3,5-Difluoro-2-hydroxyphenyl)ethanone

A496390-1G A496390-250MG A496390-25G A496390-5G A496390-10G
76774-456EA 12.74 USD
76774-456 76774-458 77551-506 77594-554 77602-140
3',5'-Difluoro-2'-hydroxyacetophenone 98%
3',5'-Difluoro-2'-hydroxyacetophenone

Precaución: for Research Use Only

Formula: C₈H₆F₂O₂
MW: 172.13 g/mol
MDL Number: MFCD00042476
CAS Number: 140675-42-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR