5-Bromo-1,4-benzodioxane 96%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-2,3-dihydro-1,4-benzodioxane

A492443-100MG A492443-10G A492443-1G A492443-250MG A492443-5G
76774-444EA 20 USD
76774-444 76774-446 76774-448 76774-450 76774-452
5-Bromo-1,4-benzodioxane 96%
5-Bromo-1,4-benzodioxane

Precaución: for Research Use Only

Formula: C₈H₇BrO₂
MW: 215.05 g/mol
MDL Number: MFCD03095029
CAS Number: 58328-39-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR