5-Amino-3-iodo-1H-indazole 95%

Supplier: Ambeed
Warning

A488034-1G A488034-250MG
76774-426EA 25 USD
76774-426 76774-428
5-Amino-3-iodo-1H-indazole 95%
5-Amino-3-iodo-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₆IN₃
MDL Number: MFCD07781656
CAS Number: 599183-36-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR