5-Chloro-2-trifluoromethylpyridine 98%
Supplier: Ambeed
|
Warning
|
A486572-10G
A486572-1G
A486572-25G
A486572-5G
76774-410EA
74.27
USD
76774-410
76774-412
76774-414
76774-416
5-Chloro-2-trifluoromethylpyridine 98%
5-Chloro-2-trifluoromethylpyridine
Precaución: for Research Use Only
Formula:
C6H3ClF3N MW: 181.5400 g/mol |
MDL Number:
MFCD06657685 CAS Number: 349-94-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...