(S)-6-(((Allyloxy)carbonyl)amino)-2-aminohexanoic acid hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A483657-1G
A483657-250MG
A483657-5G
76774-396EA
108.22
USD
76774-396
76774-398
76774-400
(S)-6-(((Allyloxy)carbonyl)amino)-2-aminohexanoic acid hydrochloride 97%
(S)-6-(((Allyloxy)carbonyl)amino)-2-aminohexanoic acid hydrochloride
Precaución: for Research Use Only
Formula:
C₁₀H₁₉ClN₂O₄ MW: 266.72 g/mol |
MDL Number:
MFCD09749825 CAS Number: 147529-99-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...