4-Bromo-N-tert-butylbenzamide 98%

Supplier: Ambeed
Warning

A482793-1G A482793-5G
76774-380EA 7 USD
76774-380 76774-382
4-Bromo-N-tert-butylbenzamide 98%
4-Bromo-N-tert-butylbenzamide

Precaución: for Research Use Only

Formula: C₁₁H₁₄BrNO
MDL Number: MFCD00462539
CAS Number: 42498-38-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR